missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone Related Compound J Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013819
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013819 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013819 Supplier Merck Emd Millipore Supplier No. PHR184630MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 28416-82-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H26F2O5
Synonym 6 alpha, 9 alpha-Difluoro-11 beta,17 alpha-dihydroxy-16 alpha-methyl-3-oxoandrosta-1,4-diene-17 beta-carboxylic acid; 6 ALPHA, 9 ALPHA-DIFLUORO-11 BETA,17 ALPHA-DIHYDROXY-16 ALPHA-METHYL-3-OXOANDROSTA-1,4-DIENE-17 BETA-CARBOXYLIC ACID
Type Certified Reference Material
Quantity 30 mg
Formula Weight 396.42
Grade Certified Reference Material