missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluticasone propionate, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 272°C to 273°C |
| Quantity | 500 mg |
| Solubility Information | Solubility in water: .. Other solubilities: soluble in dioxane |
| Formula Weight | 500.57 |
| Chemical Name or Material | Fluticasone propionate |
RUO – Research Use Only