missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoSpheres™ Carboxylate-Modified Microspheres

Achieve the brightest fluorescence with Carboxylate-Modified FluoSphere Microspheres, available in different colors and particle sizes.

$646.00 - $1,080.00

Specifications

For Use With (Application) Fluorescence Microscopy
Material Polystyrene
Product Type Carboxylate-Modified Microsphere
Product Line FLUOSPHERES
Surface Modification Carboxylate
View More Specs

Products 28
Catalog Number Mfr. No. Diameter (Metric) Color Excitation/Emission Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Color Excitation/Emission Quantity Price Quantity & Availability  
F8803
View Documents Promotion Details
Invitrogen™
F8803
0.1 μm Yellow-Green 505/515 nm 10 mL
Each for $678.00
Only null left
 
F8816
Encompass
View Documents Promotion Details
Invitrogen™
F8816
1.0 μm Crimson 625/645 nm 2 mL
Each for $678.00
Only null left
 
F8823
Encompass
View Documents Promotion Details
Invitrogen™
F8823
1.0 μm Yellow-Green 505/515 nm 10 mL
Each for $716.00
Only null left
 
F8801
View Documents Promotion Details
Invitrogen™
F8801
0.1 μm Red 580/605 nm 10 mL
Each for $678.00
Only null left
 
F8811
View Documents Promotion Details
Invitrogen™
F8811*
0.2 μm Yellow-Green 505/515 nm 10 mL
Each for $688.00
Only null left
 
F8807
Encompass
View Documents Promotion Details
Invitrogen™
F8807
0.2 μm Dark Red 660/680 nm 2 mL
Each for $684.00
Only null left
 
F10720
Encompass
View Documents Promotion Details
Invitrogen™
F10720
0.04 μm Yellow-Green, Orange, Red, Dark Red 505/515, 540/560, 580/605, 660/680 nm 1 mL/each
Each for $1,080.00
Only null left
 
F8786
View Documents Promotion Details
Invitrogen™
F8786
0.02 μm Red 580/605 nm 10 mL
Each for $676.00
Only null left
 
F8795
View Documents Promotion Details
Invitrogen™
F8795
0.04 μm Yellow-Green 505/515 nm 1 mL
Each for $674.00
Only null left
 
F8813
Encompass
View Documents Promotion Details
Invitrogen™
F8813
0.5 μm Yellow-Green 505/515 nm 10 mL
Each for $682.00
Only null left
 
F8820
Encompass
View Documents Promotion Details
Invitrogen™
F8820
1.0 μm Orange 540/560 nm 10 mL
Each for $668.00
Only null left
 
F8825
Encompass
View Documents Promotion Details
Invitrogen™
F8825
2.0 μm Nile Red 535/575 nm 2 mL
Each for $654.00
Only null left
 
F8782
Encompass
View Documents Promotion Details
Invitrogen™
F8782
0.02 μm Crimson 625/645 nm 2 mL
Each for $666.00
Only null left
 
F8784
Encompass
View Documents Promotion Details
Invitrogen™
F8784
0.02 μm Nile Red 535/575 nm 10 mL
Each for $666.00
Only null left
 
F8787
View Documents Promotion Details
Invitrogen™
F8787
0.02 μm Yellow-Green 505/515 nm 10 mL
Each for $668.00
Only null left
 
F8792
Encompass
View Documents Promotion Details
Invitrogen™
F8792
0.04 μm Orange 540/560 nm 1 mL
Each for $662.00
Only null left
 
F8793
Encompass
View Documents Promotion Details
Invitrogen™
F8793
0.04 μm Red 580/605 nm 1 mL
Each for $688.00
Only null left
 
F8794
Encompass
View Documents Promotion Details
Invitrogen™
F8794
0.04 μm Red-Orange 565/580 nm 1 mL
Each for $662.00
Only null left
 
F8797
Encompass
View Documents Promotion Details
Invitrogen™
F8797
0.1 μm Blue 350/440 nm 10 mL
Each for $674.00
Only null left
 
F8799
Encompass
View Documents Promotion Details
Invitrogen™
F8799
0.1 μm Infrared 715/755 nm 1 mL
Each for $662.00
Only null left
 
F8800
Encompass
View Documents Promotion Details
Invitrogen™
F8800
0.1 μm Orange 540/560 nm 10 mL
Each for $656.00
Only null left
 
F8805
Encompass
View Documents Promotion Details
Invitrogen™
F8805
0.2 μm Blue 365/415 nm 10 mL
Each for $660.00
Only null left
 
F8806
Encompass
View Documents Promotion Details
Invitrogen™
F8806
0.2 μm Crimson 625/645 nm 2 mL
Each for $678.00
Only null left
 
F8814
Encompass
View Documents Promotion Details
Invitrogen™
F8814
1.0 μm Blue 365/415 nm 10 mL
Each for $662.00
Only null left
 
F8815
Encompass
View Documents Promotion Details
Invitrogen™
F8815
1.0 μm Blue 350/440 nm 10 mL
Each for $646.00
Only null left
 
F8819
Encompass
View Documents Promotion Details
Invitrogen™
F8819
1.0 μm Nile Red 535/575 nm 10 mL
Each for $674.00
Only null left
 
361055995
View Documents Promotion Details
Invitrogen™
F8821
1.0 μm Red 580/605 nm 10 mL
Each for $676.00
Only null left
 
F8824
Encompass
View Documents Promotion Details
Invitrogen™
F8824
2.0 μm Blue 365/415 nm 2 mL
Each for $658.00
Only null left
 
Description

Description

Easily perform flow cytometry, microscopy, HTS, HCS, immunoassay, and other laboratory applications using our extensive selection of FluoSpheres Carboxylate-Modified Microspheres. FluoSphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications. FluoSphere microspheres are loaded with proprietary fluorescent dyes, making them the brightest microspheres available.

Visualize the brightest fluorescence for laboratoy applications including fluorescence microscopy, flow cytometry, HTS, HCS, and cell tracing with our Carboxylate-Modified FluoSphere Microspheres, which are manufactured from polystyrene microspheres and loaded with different proprietary dyes. Using specialized staining methods enables all of the fluorescent dye molecules to be contained inside each polystyrene microsphere instead of on the bead's surface. This protective environment within the bead shields the dye from detrimental environmental effects, such as photobleaching. Our carboxylate-modified microspheres are coated with a hydrophilic polymer containing multiple carboxylic acids for covalent attachment of ligands. A range of particle sizes is available for different research uses and experiments.
Specifications

Specifications

Fluorescence Microscopy
Carboxylate-Modified Microsphere
Carboxylate
Polystyrene
FLUOSPHERES
Videos
SDS
Documents

Documents