missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoroquinolonic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 86393-33-1 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Physical Form | Neat |
| Linear Formula | C13H9 ClFNO3 |
| MDL Number | MFCD00792458 |
| Melting Point | 242°C to 245°C |
| Synonym | 7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid; Ciprofloxacin Related Compound A (USP); Ciprofloxacin impurity A (PhEur); Fluoroquinolonic acid; Q-Acid |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 281.67 |
| Show More |