missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzene, 99%

Numéro de catalogue. p-4509582
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC119341000 100 g Glass bottle
AC119345000 500 g Glass bottle
AC119340025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC119341000 Fournisseur Thermo Scientific Chemicals Code fournisseur 119341000
Il en reste null

CAS: 462-06-6 | C6H5F | 96.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 462-06-6
Molecular Formula C6H5F
Molecular Weight (g/mol) 96.10
MDL Number MFCD00000280
InChI Key PYLWMHQQBFSUBP-UHFFFAOYSA-N
Synonym benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene
PubChem CID 10008
ChEBI CHEBI:5115
IUPAC Name fluorobenzene
SMILES FC1=CC=CC=C1

Spécifications

Melting Point -42.0°C
Color Colorless
Density 1.0200g/mL
Boiling Point 85.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4643 to 1.4663
Linear Formula C6H5F
Quantity 100 g
Beilstein 05,IV,632
Merck Index 15,4201
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and water
Formula Weight 96.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Fluorobenzene
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
EINECSNumber 207-321-7

RUO – Research Use Only