missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoro-Max Fluorescent Streptavidin-Coated Particles

Catalog No. 09981112
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09981112 5 mL
09981111 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09981112 Supplier Thermo Scientific™ Supplier No. 29470701010150

Simplify the binding process with a universal binding reagent.

Simplify the binding process with a universal binding reagent by using Thermo Scientific™ Fluoro-Max Fluorescent Streptavidin-Coated Particles in a variety of applications such as clinical diagnostics, immuno/histological studies and research. With an extremely broad Stokes shift, europium chelate particles help prevent nonspecific fluorescence interference. They are also excellent for quantitative membrane-based rapid assays, heterogeneous assays, luminescent assays, research applications, phagocytosis studies, cell surface markers and pore size determination.

When excited with UV light at its maximal absorbance wavelength of 333 nm, europium chelate particles emit long-lived luminescence at 613 nm, with a lifetime of approximately 0.5 milliseconds, which is about 10,000 to 100,000 times longer than the lifetimes of most fluorophores. This extremely long lifetime allows the application of europium particles to time-resolved fluorescence and can reduce the detection limits of fluorescence assays many orders of magnitude by elimination of background interference from relatively short-lived matrix fluorescence.
  • Made by dyeing Thermo Scientific™ OptiLink™ Carboxylate-Modified particles with europium chelate
  • Specifically designed for membrane or automated fluorometric-based applications
  • Available in standard 0.1 μM, 0.2 μM, and 0.3 μM diameters
  • Dyed internally to prevent dye leaching and to assure maximum surface immunoreactivity

Fluoro-Max Fluorescent Benefits

  • Bind ligands without dye interference
  • Dye-free surface for coupling
  • High protein binding capacity
  • Readily adsorbs proteins
  • Optimize assays by controlling sensitivity, specificity and stability
  • Optimized acid content

Recommended for:

  • Clinical diagnostics
  • Molecular biology
  • Lateral flow, turbidimetric and slide aggulation assays

Specifications

Additive 0.05% Sodium Azide
Binding Property Streptavidin
Concentration 1% solids
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Contents Fluorescent red dyed polystyrene microspheres in water
Color White
For Use With (Application) Immunohistological Studies
Material Polystyrene or Polystyrene with Coploymer Grafted Surface Coated with Streptavidin
Product Type Fluorescent Streptavidin-Coated Particles
Dye Type Europium Chelate
Includes Package Insert Sheet containing Certicate of Analysis
Parking Area Biotin binding of ∼1000pmol/mg
Particle Density 1.06g/cm³
Quantity 5 mL
Stability Product stable for a minimum of 24 months
Surface Modification Streptavidin
Uniformity <5% CV
Diameter (Metric) Mean 0.3 μm
Diameter (Metric) Nominal 0.3 μm
Diameter (Metric) 0.3 μm
Show More Show Less