missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, Certified, 5.00ppm ±0.10ppm (1mL = 0.005mg F-), LabChem™

Catalog No. LC145651
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC145651 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC145651 Supplier LabChem Supplier No. LC145651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-49-4 , 7732-18-5
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Poly Bottle
CAS Max % 99.999, 0.001
Identification Passes Test
Physical Form Liquid
Concentration 5.00 ppm ±0.10 ppm (1mL = 0.005mg F-)
Linear Formula NaF
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 41.99
Grade Certified
Chemical Name or Material Fluoride Standard, 5ppm (1mL = 0.005mg F-)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature