missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride Standard, 10 ppm F-, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Odor | Odorless |
| Assay Percent Range | 10.00 ± 0.05ppm |
| Boiling Point | 100°C |
| Solubility | Infinitely soluble in water |
| Solvent or Matrix | Water |
| Quantity | 4 L |
| Chemical Name or Material | Fluoride Standard, 1mL = 0.01mg F-, 10ppm F- |