missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 10 ppm F-, Ricca Chemical

Catalog No. R3170000120
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R3170000120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3170000120 Supplier Ricca Chemical Company Supplier No. R3170000120A
Only null left

Specifications

Packaging Natural Poly Bottle
CAS 7681-49-4, 7732-18-5
Odor Odorless
Assay Percent Range 10.00 ± 0.05ppm
Molecular Formula FNa
MDL Number MFCD00003524
Solubility Infinitely soluble in water
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES [F-].[Na+]
Solvent or Matrix Water
IUPAC Name sodium fluoride
Molecular Weight (g/mol) 41.99
Quantity 120 mL
PubChem CID 5235
ChEBI CHEBI:28741
Chemical Name or Material Fluoride Standard, 1mL = 0.01mg F-, 10ppm F-
Show More Show Less