missing translation for 'onlineSavingsMsg'
Learn More

Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL

Catalog No. AA35550AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35550AE 100 mL
AA35550AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35550AE Supplier Thermo Scientific Chemicals Supplier No. 035550AE

CAS: 7681-49-4 | FNa | 41.99 g/mol

Fluoride, Ion chromatography standard solution, Specpure™, F∣- 1000μg/ml is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-49-4 , 7732-18-5
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium;fluoride
SMILES [F-].[Na+]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Solubility Immiscible with water
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Sodium fluoride: 0.22%; Water: 99.78%
Chemical Name or Material Fluoride
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only