missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Fluorescin [Reagent for Oxydases and Peroxydases] | |
| 127°C | |
| MFCD00046931 | |
| MURGITYSBWUQTI-UHFFFAOYSA-N | |
| 2-(3,6-dihydroxy-9H-xanthen-9-yl)benzoic acid | |
| 68205 | |
| 334.33 | |
| Crystalline Powder |
| 518-44-5 | |
| C20H14O5 | |
| 25 g | |
| C1=CC=C(C(=C1)C2C3=C(C=C(C=C3)O)OC4=C2C=CC(=C4)O)C(=O)O | |
| 334.327 | |
| CHEBI:42492 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 518-44-5 | |
| 334.327 | |
| MURGITYSBWUQTI-UHFFFAOYSA-N | |
| CHEBI:42492 | |
| C1=CC=C(C(=C1)C2C3=C(C=C(C=C3)O)OC4=C2C=CC(=C4)O)C(=O)O |
| C20H14O5 | |
| MFCD00046931 | |
| 68205 | |
| 2-(3,6-dihydroxy-9H-xanthen-9-yl)benzoic acid |
Safety and Handling
TSCA : No