missing translation for 'onlineSavingsMsg'
Learn More

Fluorescin 95.0+%, TCI America™

Catalog No. p-7129448
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00285G 5 g
F002825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00285G Supplier TCI America Supplier No. F00285G
Only null left

Chemical Identifiers

CAS 518-44-5
Molecular Formula C20H14O5
Molecular Weight (g/mol) 334.327
MDL Number MFCD00046931
InChI Key MURGITYSBWUQTI-UHFFFAOYSA-N
PubChem CID 68205
ChEBI CHEBI:42492
IUPAC Name 2-(3,6-dihydroxy-9H-xanthen-9-yl)benzoic acid
SMILES C1=CC=C(C(=C1)C2C3=C(C=C(C=C3)O)OC4=C2C=CC(=C4)O)C(=O)O

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 334.33
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescin [Reagent for Oxydases and Peroxydases]