missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein diacetate, 97%, pure

Numéro de catalogue. AC191660250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC191660250 25 g Glass bottle
AC191660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC191660250 Fournisseur Thermo Scientific Chemicals Code fournisseur 191660250
Il en reste null

CAS: 596-09-8 | C24H16O7 | 416.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 596-09-8
Molecular Formula C24H16O7
Molecular Weight (g/mol) 416.39
MDL Number MFCD00005062
InChI Key CHADEQDQBURGHL-UHFFFAOYSA-N
Synonym fluorescein diacetate, 3,6-diacetoxyfluoran, diacetylfluorescein, di-o-acetylfluorescein, fluorescein, diacetate, 3',6'-diacetylfluorescein, unii-yl39r93pre, yl39r93pre, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 3',6'-bis acetyloxy, 3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-3',6'-diyl diacetate
PubChem CID 65047
IUPAC Name (6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3

Spécifications

Melting Point 200.0°C to 205.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 19,227
Formula Weight 416.39
Percent Purity 97%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein diacetate
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 209-877-6

RUO – Research Use Only