missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Chloride 95.0+%, TCI America™

Catalog No. D036925G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D036925G 25 g
D03691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D036925G Supplier TCI America Supplier No. D036925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 630-88-6
Molecular Formula C20H10Cl2O3
Molecular Weight (g/mol) 369.20
MDL Number MFCD00037841
InChI Key LNBKJWCRFAZVLK-UHFFFAOYSA-N
PubChem CID 99508
IUPAC Name 3',6'-dichloro-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES ClC1=CC=C2C(OC3=CC(Cl)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1

Specifications

Quantity 25 g
Synonym 3′,6′-Dichlorofluoran
Formula Weight 369.20
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein Chloride [Reagent for Amines]
EINECSNumber (5)-0657
TSCA Yes