missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein

Catalog No. AAL1325122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1325122 100 g
AAL1325136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1325122 Supplier Thermo Scientific Chemicals Supplier No. L1325122

CAS: 2321-07-5 | C20H12O5 | 332.31 g/mol

Fluorescein is used as a fluorescent tracer for many applications including in a type of dye laser as the gain medium, in forensics and serology to detect latent blood stains and in dye tracing. It is used to localise multiple muscular ventricular septal defects during open heart surgery and confirm the presence of any residual defects. It is applied to teeth to reveal plaque.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water and alcohol.

Notes
Incompatibles with strong oxidizing agents. Moisture sensitive.

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
Synonym fluorescein, solvent yellow 94, resorcinolphthalein, yellow fluorescein, 3,6-fluorandiol, d and c yellow no. 7, fluoresceine, japan yellow 201, c.i. solvent yellow 94, d&c yellow no. 7
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 320°C
Quantity 100 g
Beilstein 94324
Merck Index 14,4159
Solubility Information Slightly soluble in water and alcohol.
Formula Weight 332.32
Percent Purity ≥90%
Chemical Name or Material Fluorescein
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 219-031-8
RTECSNumber LM5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only