missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluocinolone Acetonide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 266°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 452.49 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Fluocinolone Acetonide |
Safety and Handling
| RTECSNumber | TU3830000 |
| TSCA | No |