missing translation for 'onlineSavingsMsg'
Learn More

Flunixin Meglumine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014179
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014179 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014179 Supplier Merck Emd Millipore Supplier No. PHR14421G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flunixin meglumine is a nonsteroidal anti-inflammatory drug and a potent cyclo-oxygenase (COX) inhibitor. It is commonly used as an analgesic and antipyretic in animals.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 42461-84-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C14H11F3N2O2·C7H17 NO5
MDL Number MFCD01725419
Synonym Flunixin meglumine; 2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid meglumine salt; Banamine
Type Certified Reference Material
Quantity 1 g
Formula Weight 491.46
Grade Certified Reference Material