missing translation for 'onlineSavingsMsg'
Learn More

Flumetasone 98.0+%, TCI America™

Catalog No. F0945200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0945200MG 200 mg
F09451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0945200MG Supplier TCI America Supplier No. F0945200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2135-17-3
Molecular Formula C22H28F2O5
Molecular Weight (g/mol) 410.458
MDL Number MFCD00056464
InChI Key WXURHACBFYSXBI-GQKYHHCASA-N
PubChem CID 16490
ChEBI CHEBI:34764
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES CC1CC2C3CC(C4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C)F

Specifications

Melting Point 244°C
Color White
Quantity 200 mg
Formula Weight 410.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Flumetasone
EINECSNumber (9)-2154
RTECSNumber TU3832000
TSCA No
Recommended Storage Refrigerator