Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Supplier:  Thermo Scientific™ SH31118.02
 Afficher plus de versions de ce produit
Numéro de catalogue. SH3111802
Il en reste null
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
  • Irradiation dose of 25-40 kGy provides minimum SAL of 10-6 for product contact surfaces
Spécifications
Identifiants chimiques
Extension Set
Spécifications
1/4 in. x 7/16 in. OD
MPC
TPE 374, 2.5 m (8.2 ft)
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 2.5 m (8.2 ft); MPC
Extension Set
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS