Learn More
Thermo Scientific™ Fluid Transfer Assembly
GREENER_CHOICE
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Supplier:  Thermo Scientific™ SS00064I
 Afficher plus de versions de ce produit
Numéro de catalogue. SS00064I
Il en reste null
Description
Description
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.
  • End treatments facilitate simple interconnection with existing process and equipment
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
Spécifications
Identifiants chimiques
Fluid Transfer Assembly
Spécifications
91.4cm of 12.7cm ID Silicone Tubing with 3.8cm Sanitary and 1.3cm MPX insert Connections
3.8 cm Sanitary/MPX Insert
Fluid Transfer Assembly
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS