missing translation for 'onlineSavingsMsg'
Learn More

Flucytosine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111013458
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013458 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013458 Supplier Merck Emd Millipore Supplier No. PHR16591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Flucytosine is an antimycotic drug that can be effectively utilized against severe Candida and Cryptococcus infections.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 2022-85-7
Linear Formula C4H4FN3O
MDL Number MFCD00006035
Melting Point 298°C to 300°C (decomposition)
Synonym 5-Fluorocytosine; 4-amino-5-fluoro-2(1 H)-pyrimidinone; Flucytosine
Quantity 1 g
Formula Weight 129.09
Grade Certified Reference Material