missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluconazole Related Compound C, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C10H8N6 |
| Synonym | 1,1′-(1,3-Phenylene)bis-1 H-1,2,4-triazole; 1,1′-(1,3-Phenylene)-di-(1 H-1,2,4-Triazole) |
| Type | Certified Reference Material |
| Quantity | 20 mg |
| Formula Weight | 212.21 |
| Grade | Certified Reference Material |