missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015614
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015614 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015614 Supplier Merck Emd Millipore Supplier No. PHR186220MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 81886-51-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C13H13 FN6 O
Synonym 2-(4-Fluorophenyl)-1,3-bis(1 H-1,2,4-triazol-1-yl)propan-2-ol; 2-(4-fluorophenyl)-1,3-bis(1 H-1,2,4-triazol-1-yl)-propan-2-ol
Type Certified Reference Material
Quantity 20 mg
Formula Weight 288.28
Grade Certified Reference Material