Chemglass Inc Low Form Dewar Flask, Hemispherical, 600mL Capacity, Fits 500mL RBF, 140mm ID X 65mm Inside Depth, 165mm OD X 95mm Height
Numéro de catalogue. CG159003
Low Form Dewar Flask, Hemispherical, 600mL Capacity, Fits 500mL RBF, 140mm ID X 65mm Inside Depth, 165mm OD X 95mm Height. Low form hemispherical Dewar with protective aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Numéro de catalogue.
CG159003
Fournisseur
Chemglass IncCode fournisseur
CG159003
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Low Form Dewar Flask, Hemispherical, 600mL Capacity, Fits 500mL RBF, 140mm ID X 65mm Inside Depth, 165mm OD X 95mm Height. Low form hemispherical Dewar with protective aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.