missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Florisil™ PR Grade (analysis acc. to FDA)

Catalog No. 111012975
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012975 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012975 Supplier MilliporeSigma™ Supelco™ Supplier No. 46382500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Florisil™ was used: as column packing material in column chromatography. as an adsorbent in gas chromatography with electron capture detector (GC/ECD), capillary column chromatography, adsorption column chromatography. in clean-up procedures in gas chromatography-tandem mass spectrometry and GC-FID

Florisil™, adsorbent for the separation of lipids, is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil™ is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil™ cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Specifications

Pore Size 60 Å
Grade PR Grade (analysis acc. to FDA)
Quantity 500 g
Surface Area (Metric) 289 m2/g
Product Line Florisil
Physical Form Coarse Powder
MDL Number MFCD00078249
CAS 1343-88-0

Florisil is a registered trademark of U.S. Silica Company