missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, Activated at 675°C, 60 to 100 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003124
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003124 500 g Wide Mouth Amber Glass Bottle
02003123 2 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003124 Supplier Avantor Supplier No. 337207

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

    Suitable for Use in Chromatographic Cleanup of Pesticide Residues

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Quantity 500 g
Mesh Size 60 to 100 Mesh
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.51kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Florisil™, Activated at 675°C