missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, 60 to 100 Mesh, J.T. Baker™

Catalog No. 02003122
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003122 500 g Poly Bottle
02003118 2 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003122 Supplier Avantor Supplier No. M36807

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Selective Adsorbent for Gas Chromatography

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Quantity 500 g
Mesh Size 60 to 100 Mesh
Packaging Poly Bottle
Specific Gravity 1L = 2.51kg
Chemical Name or Material Florisil™