missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Flavin adenine dinucleotide disodium salt
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Yellow to Orange |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 250 mg |
| Solubility Information | Solubility in water: 5g/L |
| Spectrophotometry | 90% (lambda min. 450nm, pH 7), molar absorptivity 11300 +/- 4%, on dry substance |
| Formula Weight | 829.52 |
| Physical Form | Powder |
| Chemical Name or Material | Flavin adenine dinucleotide disodium salt |
Sécurité et manipulation
| EINECSNumber | 282-733-8 |
RUO – Research Use Only