Learn More
BeLLCo Glass Stainless steel bottom base for extra strength includes four rubber feet for added stability. Carriers are dishwasher and oven safe. For use with 6L or 9L vessels or carboys. Material: stainless steel Dimensions: 9""
Numéro de catalogue. 501945982
Encompass
Stainless steel bottom base for extra strength includes four rubber feet for added stability. Carriers are dishwasher and oven safe. For use with 6L or 9L vessels or carboys. Material: stainless steel Dimensions: 9"
Numéro de catalogue. 501945982 Fournisseur BeLLCo Glass Code fournisseur 766906000
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Stainless steel bottom base for extra strength includes four rubber feet for added stability. Carriers are dishwasher and oven safe. For use with 6L or 9L vessels or carboys. Material: stainless steel Dimensions: 9"