missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Silica

Catalog No. 05410008
Change view
Click to view available options
Includes:
1 Pc.
10 Pc.
12 Pc.
18 Pc.
2 Pc.
20 Pc.
3 Pc.
5 Pc.
6 Pc.
Bed Weight (Metric):
4 g
12 g
25 g
40 g
80 g
120 g
220 g
330 g
800 g
1600 g
3000 g
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Bed Weight (Metric)
05410008 10 Pc. 120 g
05410004 18 Pc. 12 g
05410012 2 Pc. 1600 g
05410009 6 Pc. 220 g
05410005 12 Pc. 25 g
05410013 1 Pc. 3000 g
05410010 5 Pc. 330 g
05410006 12 Pc. 40 g
05410003 20 Pc. 4 g
05410011 3 Pc. 800 g
05410007 10 Pc. 80 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 05410008 Supplier BUCHI Supplier No. 140000026

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.

  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters

Specifications

Material Polypropylene
Type Silica
Includes 10 Pc.
Resolution 40 to 63 μm
Bed Weight (Metric) 120 g
Column Type Silica
For Use With (Application) Basic Applications, Polar Compounds
For Use With (Equipment) Any Flash System