missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 660

Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.

$197.00 - $594.00

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
501128848
View Documents Promotion Details
Invitrogen™
65086414
100 tests
Each for $197.00
Only null left
 
5016964
View Documents Promotion Details
Invitrogen™
65086418
500 tests
Each for $594.00
Only null left
 
Description

Description

  • Fixable Viability Dyes may be used to label cells from all species
  • Fixable Viability Dye eFluor™ 660 can be excited by the red (633nm) laser line and has a peak emission of 660nm that can be detected using a 660/20 band pass filter (equivalent to APC or Alexa Fluor™ 647)
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute. After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye.
  • Fixable Viability Dye eFluor™ 660 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture
  • Store at −70°C with dessicant
  • It may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
  • Staining may be done before or after surface staining
  • Cells may be cryopreserved after staining with no adverse effect on staining intensity of dead cells after thawing
Specifications

Specifications

Red
Viability Assay
Dye
eBioscience
Fluorescence
Flow Cytometer
633/660
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.