missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 660

Catalog No. 501128848
Change view
Click to view available options
Quantity:
100 tests
500 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501128848 100 tests
5016964 500 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501128848 Supplier Invitrogen™ Supplier No. 65086414

Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.

  • Fixable Viability Dyes may be used to label cells from all species
  • Fixable Viability Dye eFluor™ 660 can be excited by the red (633nm) laser line and has a peak emission of 660nm
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • Fixable Viability Dye eFluor™ 660 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture
  • Store at −70°C with dessicant
  • It may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
  • Staining may be done before or after surface staining
  • Cells may be cryopreserved after staining with no adverse effect on staining intensity of dead cells after thawing

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
Emission 633/660
Product Line eBioscience
Quantity 100 tests

For Research Use Only. Not for use in diagnostic procedures.