Learn More
ACROBiosystems Inc FITC-Labeled Human CD47 Protein, His Tag
Numéro de catalogue. 502054743
Encompass
Recombinant Protein;25UG;FITC-Labeled Human CD47, His Tag (CD7-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa.
Numéro de catalogue. 502054743 Fournisseur ACROBiosystems Inc Code fournisseur CD7HF2H325UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;25UG;FITC-Labeled Human CD47, His Tag (CD7-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa.