Learn More
Fisherbrand™ USB Data Logger Interface for Micro Series Data Loggers
Catalog No. 15060037
Change view
Click to view available options
For Use With (Equipment):
Micro series dataloggers
Catalog No. For Use With (Equipment)
15060037 Micro series dataloggers
1 options
Catalog No. 15060037 Supplier Fisherbrand™ Supplier No. IFC202
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
USB interface cable with 3.5mm headphone plug and 3.5mm to 2.5mm adapter
IFC202 interface cable communicates between the Micro series data loggers and the MadgeTech software. Through this interface, loggers can be started, stopped or downloaded. Pllug and play: it can be directly connected to a computer without installing any drivers and is compatible with Windows XP SP3, Windows Vista (32 and 64 bit), and Windows 7 (32 and 64 bit) operating systems. Can operate at up to 500 Volts RMS relative to the computers earth ground when attached. Communication LEDs provide a quick indication of the device status. The blue light illuminates when the device is plugged in and successfully recognized by Windows. The red light flashes when data is sent, and the green light flashes when data is received.
  • For communicating with 15-060-006 and 15-060-007
  • Blue, green and red LED indicators
  • Reduced noise with lionger sensor leads
  • Plug and play; no driver installation required
Specifications
Type USB Interface Cable Package
Includes Cable with 3.5mm headphone plug, 3.5mm to 2.5mm adapter
For Use With (Equipment) Micro series dataloggers