missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Standard Disposable Transfer Pipettes
GREENER_CHOICE

Catalog No. 137119D
Change view
Click to view available options
Volume (Metric):
7.5 mL
Graduated:
Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Graduated
137119D 7.5 mL Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 137119D Supplier Thermo Scientific™ Supplier No. 225
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single squeeze draws up to 3.2 mL

Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials.

  • Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel
  • One-piece design molded from see-through, low-density polyethylene
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • In four ready-to-use styles
  • One squeeze draws from 2.0 to 3.3 mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)

Specifications

Bulb Draw (Metric) 3.2 mL
Material Polyethylene
For Use With (Application) Research
Graduated Yes
Quantity 500/Pk.
Volume (Metric) 7.5 mL
Length (Metric) 15.5 cm
Type Transfer Pipet
Disposable Single-use