missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MicroMat™ CLR Silicone Mats for Well Plates

Catalog No. 0339652
Change view
Click to view available options
Plate Compatibility:
96-well Plate
Well Shape:
Round
Pre-slit:
No
Yes
Includes:
Large Penetration Area
Small Penetration Area
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility Well Shape Pre-slit Includes
0339652 96-well Plate Round Yes Small Penetration Area
0339649 96-well Plate Round No Small Penetration Area
0339650 96-well Plate Round Yes Large Penetration Area
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0339652 Supplier Fisherbrand™ Supplier No. 0339652

Safely seal plates and eliminate cross-contamination of samples with Fisherbrand™ MicroMat™ CLR Silicone Mats for Well Plates. Manufactured of pure silicone these mats resist coring and tearing and provide superior resealability after multiple injections.

Excellent Chemical and Thermal Compatibility

  • Solvent-resistant with excellent chemical compatibility
  • Dry-heat autoclavable and can withstand low temperatures to -80°C

Automation-Ready

  • Footprint allows plates to be securely stacked without robotic arm interference
  • Pre-slit mats offer better piercability and reduced vacuum formation

Specifications

Color Clear
Solvent Or Alcohol Resistant No Resistance
Material Silicone
Autoclavable Autoclavable
No. of Wells 96
Includes Small Penetration Area
No. per Pack 10
Plug Diameter 7 mm
Pre-slit Yes
Product Type Sealing Mat
Product Line Fisherbrand
Temperature Range (Metric) -80°C
Well Shape Round
Plate Compatibility 96-well Plate
Show More Show Less