missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Radioactive Indicator Dots
GREENER_CHOICE

Catalog No. 18999999
Change view
Click to view available options
Format:
Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
18999999 Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999999

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Radiation Indicator dots adhere to laboratory glassware and equipment to identify caution.

  • The labels can be printed on and will not bleed.
  • Perfect for distinguishing amongst various specimens.
Note: These dots are for labeling a radioactive substance. They do not change color or indicate if something has gone through gamma radiation for bioreduction or sterility. 
 

Specifications

Description Radioactive Indicator Dots
Format Roll
For Use With (Application) Place these dots on equipment or lab items to identify a radioactive substance.
Includes 1 roll (1000 dots)
Shape Round