Learn More
Fisherbrand™ Polystyrene Beakers with Pouring Lip
Catalog No. 14955112
Change view
Click to view available options
Capacity (Metric):
190 mL
Catalog No. Capacity (Metric)
14955112 190 mL
1 options
Catalog No. 14955112
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For urine or sputum
  • Clear polystyrene
  • Top-to-bottom channel for “Quantity Not Sufficient” specimen and strip tests
  • Resistant to dilute or weak acids
  • Stackable
  • Graduated in oz., cc and mL
Specifications
Color Clear
Capacity (Metric) 190 mL
Material Polystyrene
Includes Pouring Lip
Type Beaker
Disposable Single-use
Graduated Yes