missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ NFR Self-Laminating Labels
GREENER_CHOICE

Catalog No. 18999791
Change view
Click to view available options
Width (English):
3.5 in.
5 in.
Height (English):
2 in.
3 in.
Height (Metric):
5 cm
8 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Height (English) Height (Metric)
18999791 5 in. 3 in. 8 cm
18999790 3.5 in. 2 in. 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18999791 Supplier Fisherbrand™ Supplier No. 22681

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use on bottles, drums or any containers holding hazardous substances.

  • Each self-adhesive label features a convenient laminate cover and space to fill in National Fire Protection Association (NFPA) hazard ratings
  • Add the appropriate hazard rating number and circle the symbols of personal protection required
  • Peel off backing to apply

Specifications

Format Label
Product Type NFR Self-Laminating Label
For Use With (Application) Bottles, drums or any containers holding hazardous substances
Quantity 25/Pk.
Width (English) 5 in.
Height (English) 3 in.
Height (Metric) 8 cm