missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Large Biohazard Labels
GREENER_CHOICE

Catalog No. 18999936
Change view
Click to view available options
Quantity:
320/Pk.
640/Pk.
Size (English):
0.5 x 1.5 in.
3 x 3 in.
Size (Metric):
1.3 x 3.8 cm
7.6 x 7.6 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size (English) Size (Metric)
18999936 320/Pk. 3 x 3 in. 7.6 x 7.6 cm
18999935 640/Pk. 0.5 x 1.5 in. 1.3 x 3.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18999936 Supplier Fisherbrand™ Supplier No. LBH 24
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

Strong adhesive ensures that labels remain in place.

Compliance

Meet OSHA requirements.

Specifications

Certifications/Compliance OSHA
Color Black, Red
Format Label
Material Paper
Product Type Biohazard Warning Tape
For Use With (Application) Biohazard
Quantity 320/Pk.
Size (English) 3 x 3 in.
Size (Metric) 7.6 x 7.6 cm
Dimensions (L x W) 3 in. x 3 in.