Learn More
Fisherbrand™ Interface Cable Package for RFOT and Therm-A-Lert Data Loggers
Catalog No. 15059502
Change view
Click to view available options
Includes:
Software CD, software manual, Quick Start Guide, USB to mini USB adapter and the wireless receiver
Catalog No. Includes
15059502 Software CD, software manual, Quick Start Guide, USB to mini USB adapter and the wireless receiver
1 options
Catalog No. 15059502 Supplier Fisherbrand™ Supplier No. RFC1000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Fisherbrand™ Wireless Meat Temperature Logger (15-059-501) and Therm-A-Lert Data Loggers
Wireless transceiver for Fisher's two-way wireless dataloggers. It includes software CD, software manual, Quick Start Guide, USB to mini USB adapter and the wireless receiver.
Specifications
Type Wireless Transceiver
Includes Software CD, software manual, Quick Start Guide, USB to mini USB adapter and the wireless receiver
For Use With (Application) Used as a repeater or plugged directly into your PC
For Use With (Equipment) Wireless meat temperature logger, Therm-A-Lert dataloggers