missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ NFR Write-On Labels
GREENER_CHOICE

Catalog No. 18999789
Change view
Click to view available options
Length (English):
3.87 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18999789 3.87 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999789 Supplier Fisherbrand™ Supplier No. NEVS607

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.

  • NFR paper labels are large enough to include the chemical name, common name and manufacturer's name
  • NFR labels let you write in the chemical name, owner, date, and hazard rating

 

Order Info

Labels measure 3.87L x 1.50 in. W (9.8 x 3.8cm).

Specifications

Color Assorted
Format Label
Product Type NFR Write-On™ Label
For Use With (Application) Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers
Quantity 1000/Roll
Dimensions (L x W) 3.87 x 1.5 in.
Length (English) 3.87 in.
Length (Metric) 9.8 cm
Width (English) 1.5 in.