missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ HMIG Write-On Labels
GREENER_CHOICE

Catalog No. 18999788
Change view
Click to view available options
Length (English):
3.88 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18999788 3.88 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999788 Supplier Fisherbrand™ Supplier No. 63171

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.

  • Hazardous Materials Identification Guide (HMIG) write on labels
  • HMIG paper labels are large enough to include the chemical name, common name and manufacturer's name

Order Info

Labels measure 3.87L x 1.50 in. W (9.8 x 3.8cm).

Specifications

Color Assorted
Format Label
Product Type HMIG Write-On™ Label
For Use With (Application) Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers
Quantity 1000/Roll
Dimensions (L x W) 3.87 x 1.5 in.
Length (English) 3.88 in.
Length (Metric) 9.8 cm
Width (English) 1.5 in.