missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hellmanex™ III Liquid Cleaning Concentrate

Catalog No. 14385864
Change view
Click to view available options
Hellmanex III
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14385864 Hellmanex III
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14385864 Supplier Fisherbrand™ Supplier No. 320.003
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides effective cleaning of glass and quartz cells and other sensitive optical components

  • Highly effective alkaline concentrate
  • Contains components for trace-free decontamination without damaging high-precision polished surfaces
  • Removes fats, oils, waxes, dried blood, proteins, silicone oil, and traces of many other organic and inorganic substances
  • No influence on enzymatic or serological reactions
  • Low phosphate content

Specifications

For Use With (Application) Provides effective cleaning of glass and quartz cells and other sensitive optical components
Includes 1 Liter
Product Type Hellmanex™ III Liquid Cuvette Cleaner