missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Handheld Labeling System

Catalog No. 15915
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
15915 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15915

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy-to-use, lightweight, dating and coding system for the laboratory.

  • Handheld labeler loads quickly and dispenses labels without jamming
  • Available labels include “Expires,” “Received Opened,” “Sterile,” and blank white
  • All labels allow user to dial in two lines of information with up to eight characters per line to be printed (see inset)
  • Available characters include two Gregorian dates for printing expiration and autoclave dating with an additional band for load and sterilizer numbers
  • Contains dollar signs for price-marking applications
  • Labels have autoclavable adhesive that can withstand temperatures up to 132°C (270°F) for 20 min.

Order Info

Label size: 0.62 x 0.93 in. (1.6 x 2.4cm). 1000 labels per roll.

Specifications

Product Type Labeling System
Autoclavable Autoclavable
For Use With (Application) Laboratory
Includes Two gregorian dates for printing expiration and autoclave dating with an additional band for load and sterilizer numbers
Temperature Range (English) Up to270°F
Temperature Range (Metric) Up to 132°C