Learn More
Fisherbrand™ General-Purpose Broad-Tipped Forceps
Fisherbrand General-Purpose Broad-Tipped Forceps are perfect for cover glass handling.
Numéro de catalogue. 10300
Il en reste null
Edge
Description
Description
Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Grooved handle
  • Made from polished ASTM standard stainless-steel
  • Electronically passivated for corrosion resistance
  • Rockwell hardness (HRC): 40--45
  • Latex-free
  • Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
  • Quality control per U.S. Government specs
  • Produced under ISO 9002, GMP, cGMP guidelines
Spécifications
Spécifications
Broad tip
Polished ASTM standard stainless-steel
General purpose forceps
4.5 in.
11.4 cm
General
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS