missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cover for Glass Disposal Carton

Catalog No. 03421526
Change view
Click to view available options
Color:
White, Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03421526 White, Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03421526 Supplier Fisherbrand™ Supplier No. 132043001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Opens with a single touch.

  • Dishwasher-safe and autoclave-safe, the reusable disposal carton cover opens quickly and easily by pressing the built-in release tab—no batteries required
  • Store the disposable cardboard lid under the bottom of the carton and, when carton is full, simply remove the reusable Disposal Carton Cover, replace with the cardboard lid, and dispose of the carton in the usual fashion
  • Autoclavable at 250°F (121°C)
  • Fits any disposal carton that has a 12 x 12 in. (30.5 x 30.5 cm) opening including Fisherbrand Glass Disposal boxes S41264, 12-009-7A and 17-988-448
  • Has a smooth 4 x 4 in. (10.1 x 10.1 cm) area on top for applying labels
  • Provides a wider opening 20.5 x 24.4cm (8 x 9.5 in.) permitting safer and more sanitary deposit of wastes
  • Center panel is green to match the printing on glass disposal cartons providing fast visual recognition of the waste stream
  • Wash as needed

Specifications

Color White, Green
Autoclavable Autoclavable
For Use With (Equipment) For disposal cartons with 12 x 12 in. opening
Product Type Carton Cover