missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ ColorFrost™ Microscope Slides with Clipped Corners

Numéro de catalogue. 22037161
Change view
Click to view available options
Color:
Blue
Green
Pink
Yellow
Corner Style:
45° Clipped
Clipped
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color Corner Style
22037161 Green Clipped
22037160 Blue Clipped
22037162 Pink 45° Clipped
22037163 Yellow Clipped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 22037161 Fournisseur Fisherbrand™ Code fournisseur 6951WS44370621
Il en reste null

Durable, opaque printed surface inert to common laboratory chemicals and reagents

  • 45° clipped corners; color-coded frosted end
  • Raised printed writing surface eliminates need for paper labels and tedious relabeling of slides
  • Autoclavable

Spécifications

Length (Metric) 25 mm
Thickness (Metric) 1 mm
Width (Metric) 75 mm
Frosted Colorfrost
Color Green
Corner Style Clipped
Quantity 144/Pk.