missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Burn-up Bin™ Biohazard Waste Boxes

Catalog No. 19168430
Change view
Click to view available options
Design:
Bench model
Floor model
Dimensions (L x W x H):
8 x 8 x 10 in.
12 x 12 x 20 in.
12 x 12 x 27 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Dimensions (L x W x H)
19168430 Floor model 12 x 12 x 20 in.
120098A Floor model 12 x 12 x 27 in.
120098B Bench model 8 x 8 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19168430 Supplier Fisherbrand™ Supplier No. 797065

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Store flat, assemble in seconds.

  • Floor model box features integral handles for safety in use and transport
  • Corrugated cardboard—single use
  • 2 mil polyliner safely holds biohazard waste
  • Tight-fitting lid has flap opening to accept material; to dispose, simply close lid and transfer filled box to incinerator
  • Graphics and biohazard symbols readily identify contents, encourage proper use
  • Replacement bags are not available for the Burn-Up Bin as boxes are to be incinerated after use

Specifications

Design Floor model
Diameter (Metric) Outer 17.78 cm
Diameter (English) Outer 7 in.
Capacity (English) 11 gal.
Color White
Dimensions (L x W x H) 12 x 12 x 20 in.
Height (Metric) 50.8 cm
Length (Metric) 30 cm
Width (Metric) 30 cm
Liner Material Polyethylene
For Use With (Application) Quick identification for non-hazardous waste paper products
Width (English) 12 in.
Length (English) 12 in.
Thickness 2 mm
Height (English) 20 in.
Material Cardboard
Product Type Floor Biohazard Waste Box
Show More Show Less