missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Blood Bank Labels
Description
- For use with thermal and direct thermal printers and Misys™ software
- Peel-off labels on paper backing
- Perforations between each label
Order Info
Roll of 4525 labels.
Specifications
Specifications
| Format | Label |
| Material | Paper |